Learn More
Medchemexpress LLC HY-112256 5mg Medchemexpress, ACP-105 CAS:899821-23-9 Purity:>98%
SDP
Catalog No. 501969066
Encompass_Preferred
Medchemexpress, HY-112256 5mg ACP-105 CAS:899821-23-9 ACP-105 is an orally available, selective amd potent androgen receptor modulator (SARM), with pEC50s of 9.0 and 9.3 for AR wild type and T877A mutant, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9066 Supplier Medchemexpress LLC Supplier No. HY1122565MG
Medchemexpress, HY-112256 5mg ACP-105 CAS:899821-23-9 ACP-105 is an orally available, selective amd potent androgen receptor modulator (SARM), with pEC50s of 9.0 and 9.3 for AR wild type and T877A mutant, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.