Learn More
Enzo Life Sciences ACP (50mg). CAS: 137052-08-5
SDP
Supplier:  Enzo Life Sciences ALX430118M050
Encompass_Preferred
Cell permeable probe for the detection of intracellular oxidative stress. ACP can easily be deprotected with esterases to cyclic hydroxylamines, which are oxidized by ROS leading to ESR-detectable nitroxide radicals. Alternative name: 1-Acetoxy-3-carbamoyl-2,2,5,5-tetramethylpyrrolidine. Purity: ≤0.05% paramagnetic impurities (by EPR). Solubility: Soluble in water, methanol, 100% ethanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9210