Learn More
Medchemexpress LLC Acp-5862 | 2230757-47-6 | 98.3% | 481.51 g/mol | C26H23N7O3 | 5 MG
SDP
Catalog No. 5000222983
Encompass_Preferred
ACP-5862 is the major active, circulating metabolite of acalabrutinib. It is a pyrrolidine ring-opened small molecule that potently inhibits Bruton tyrosine kinase (BTK) with an IC50 of 5.0 nM and displays weak time-dependent inactivation of CYP3A4 and CYP2C8.
  • Potent BTK inhibition with reported IC50 of 5.0 nM.
  • Useful for metabolite profiling and pharmacology studies.
  • High reported purity (≈98.3%) suitable for research use.
  • Available in small milligram package sizes for discovery work.
  • Storage-stable under recommended powder and solvent conditions.
  • Well characterized: molecular weight 481.51 and formula C26H23N7O3.
Catalog No. 50-002-22983 Supplier Medchemexpress LLC Supplier No. HY1353345MG
ACP-5862 is the major active, circulating metabolite of acalabrutinib. It is a pyrrolidine ring-opened small molecule that potently inhibits Bruton tyrosine kinase (BTK) with an IC50 of 5.0 nM and displays weak time-dependent inactivation of CYP3A4 and CYP2C8.
  • Potent BTK inhibition with reported IC50 of 5.0 nM.
  • Useful for metabolite profiling and pharmacology studies.
  • High reported purity (≈98.3%) suitable for research use.
  • Available in small milligram package sizes for discovery work.
  • Storage-stable under recommended powder and solvent conditions.
  • Well characterized: molecular weight 481.51 and formula C26H23N7O3.