Learn More
Aviva Systems Biology Corp ACP1 Antibody - middle region (ARP42343_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42343_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACP1. Alternate names: HAAP, MGC111030, MGC3499ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-284-995