Learn More
Aviva Systems Biology Corp ACP5 Antibody - N-terminal region (ARP41471_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41471_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACP5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACP5. Alternate names: MGC117378, TRAP, SPENCDIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62830