Learn More
Aviva Systems Biology Corp ACPP Antibody - middle region (ARP41913_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41913_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACPP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACPP. Alternate names: ACP-3, ACP3, PAP, 5'-NTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58043