Learn More
Medchemexpress LLC ACPP/Prostatic Acid 5ug
SDP
Catalog No. 5000178867
Encompass_Preferred
The acid prostatic phosphatase (ACPP) protein encapsulated in virus-like particles (VLPs) is an acid tyrosine phosphatase that skillfully dephosphorylates a variety of substrates ACPP-VLP possesses lipid phosphatase activity and inactivates lysophosphatidic acid ACPP/Prostatic Acid Phosphatase Protein Mouse (HEK293 His) is the recombinant mouse-derived ACPP/Prostatic Acid Phosphatase protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78867 Supplier Medchemexpress LLC Supplier No. HYP744305UG
Add to Cart
Add to Cart
The acid prostatic phosphatase (ACPP) protein encapsulated in virus-like particles (VLPs) is an acid tyrosine phosphatase that skillfully dephosphorylates a variety of substrates ACPP-VLP possesses lipid phosphatase activity and inactivates lysophosphatidic acid ACPP/Prostatic Acid Phosphatase Protein Mouse (HEK293 His) is the recombinant mouse-derived ACPP/Prostatic Acid Phosphatase protein expressed by HEK293 with C-His labeled tag