Learn More
Waters Corp Waters Corporation Performance Maintenance Kit, ACQUITY H-Class Sample Manager FTN, Annual
Catalog No. 50791610 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit for the ACQUITY H-Class Sample Manager – Flow Through Needle contains all the Quality Parts necessary to keep the system operating at peak performance. The kit includes a syringe, needle, injection cartridge, and filters.
  • Type: Plates and racks.
  • System type: UPLC.
  • Instrument: ACQUITY.
  • Replacement schedule: Annual.
  • Component: ACQUITY UPLC Column Manager.
  • Units per package: 1 pk.
Catalog No. 50-791-610 Supplier Waters Corp Supplier No. 201000234
This Performance Maintenance Kit for the ACQUITY H-Class Sample Manager – Flow Through Needle contains all the Quality Parts necessary to keep the system operating at peak performance. The kit includes a syringe, needle, injection cartridge, and filters.
  • Type: Plates and racks.
  • System type: UPLC.
  • Instrument: ACQUITY.
  • Replacement schedule: Annual.
  • Component: ACQUITY UPLC Column Manager.
  • Units per package: 1 pk.