Learn More
Waters Corp Performance Maintenance Kit, ACQUITY I-Class Binary Solvent Manager, UPLC System
Catalog No. NC2330928 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the ACQUITY I-Class Binary Solvent Manager contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis.
  • Contains plungers, check valves, seals, and filters
  • Designed for UPLC system type
  • Compatible with H-Class system
  • Recommended for annual replacement
  • Includes one unit per package
Catalog No. NC2330928 Supplier Waters Corp Supplier No. 201000260
Add to Cart
Add to Cart
Request a Quote
The Waters™ Performance Maintenance Kit for the ACQUITY I-Class Binary Solvent Manager contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis.
  • Contains plungers, check valves, seals, and filters
  • Designed for UPLC system type
  • Compatible with H-Class system
  • Recommended for annual replacement
  • Includes one unit per package