Learn More
Waters Corp ACQUITY Premier Protein SEC Column, 250Å, 1.7 μm, 4.6 x 300 mm
Catalog No. NC2390104 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier Protein SEC 250 Å, 1.7 μm columns provide reliable protein size variant separations (10,000 to 650,000 Daltons) by synergistically combining MaxPeak Premier High Performance Surfaces and novel BEH-PEO Particle technologies. These advancements minimize undesired secondary interactions, reducing method development time and enabling generic methods for various samples.
  • Includes a vial of mAb Size Variant Standard.
  • Comes with a recommended MaxPeak Premier SEC 250Å, 2.5 μm, 4.6 x 30 mm Guard to extend column life and maintain resolution.
  • Engineered for optimal SEC separation performance in UPLC SEC applications.
Catalog No. NC2390104 Supplier Waters Corp Supplier No. 176004795
Add to Cart
Add to Cart
Request a Quote
ACQUITY Premier Protein SEC 250 Å, 1.7 μm columns provide reliable protein size variant separations (10,000 to 650,000 Daltons) by synergistically combining MaxPeak Premier High Performance Surfaces and novel BEH-PEO Particle technologies. These advancements minimize undesired secondary interactions, reducing method development time and enabling generic methods for various samples.
  • Includes a vial of mAb Size Variant Standard.
  • Comes with a recommended MaxPeak Premier SEC 250Å, 2.5 μm, 4.6 x 30 mm Guard to extend column life and maintain resolution.
  • Engineered for optimal SEC separation performance in UPLC SEC applications.