Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation ACQUITY APC AQ BEH Column, 125 Å, 2.5 μm, 4.6 mm x 150 mm

Catalog No. NC1690438 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

ACQUITY APC AQ columns provide efficient size-based separations for water soluble polymer characterization. The sorbent features patented Ethylene Bridge Hybrid (BEH) technology, ensuring a mechanically strong packed bed that resists shrinking and swelling, and facilitates mobile phase solvent switching. This column has an expected molecular weight range of 1,000 to 30,000 g/mole.

  • Pore size: 125 Å.
  • Particle size: 2.5 μm.
  • Inner diameter: 4.6 mm.
  • Length: 150 mm.
  • Separation mode: SEC/GPC.
  • Particle substrate: Hybrid.
  • pH range: 1 to 8.
  • Endcapped: No.
  • Silanol activity: Medium.
  • Particle shape: Spherical.
  • Endfitting type: Parker-style.
  • System: APC.
  • Technique: SEC.
  • Units per package: 1.

Catalog No. NC1690438 Supplier Waters Corp Supplier No. 186006980
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACQUITY APC AQ columns provide efficient size-based separations for water soluble polymer characterization. The sorbent features patented Ethylene Bridge Hybrid (BEH) technology, ensuring a mechanically strong packed bed that resists shrinking and swelling, and facilitates mobile phase solvent switching. This column has an expected molecular weight range of 1,000 to 30,000 g/mole.

  • Pore size: 125 Å.
  • Particle size: 2.5 μm.
  • Inner diameter: 4.6 mm.
  • Length: 150 mm.
  • Separation mode: SEC/GPC.
  • Particle substrate: Hybrid.
  • pH range: 1 to 8.
  • Endcapped: No.
  • Silanol activity: Medium.
  • Particle shape: Spherical.
  • Endfitting type: Parker-style.
  • System: APC.
  • Technique: SEC.
  • Units per package: 1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.