Learn More
Waters Corp Rebuild Kit, ACQUITY Accumulator Bio Pumphead, UPLC
Catalog No. 502391143 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit contains the plunger, seals, and check valve necessary to rebuild a Bio ACQUITY Accumulator Pumphead.
  • Compatible with UPLC systems.
  • Designed for replacement as required.
  • Includes one package unit.
Catalog No. 50-239-1143 Supplier Waters Corp Supplier No. 700010582
This rebuild kit contains the plunger, seals, and check valve necessary to rebuild a Bio ACQUITY Accumulator Pumphead.
  • Compatible with UPLC systems.
  • Designed for replacement as required.
  • Includes one package unit.