Learn More
Waters Corp Waters Accumulator Pumphead Rebuild Kit, UPLC, Plunger, Seals, Check Valve
Catalog No. 502039501 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit provides the necessary components to rebuild an Accumulator Pumphead. It is designed for performance maintenance of UPLC systems, compatible with UPLC H-Class System, nanoUPLC System, and 2D System. Replacement is scheduled as required.
  • Includes plunger, seals, and check valve
  • System type: UPLC
  • Compatible with UPLC H-Class System, nanoUPLC System, 2D System
  • Compatible with UPLC ISM, UPLC BSM, UPLC QSM, UPLC H-Class QSM, nanoASM, nanoBSM
  • Product type: Performance maintenance kits
  • Units per package: 1 pk
Catalog No. 50-203-9501 Supplier Waters Corp Supplier No. 700004746
This rebuild kit provides the necessary components to rebuild an Accumulator Pumphead. It is designed for performance maintenance of UPLC systems, compatible with UPLC H-Class System, nanoUPLC System, and 2D System. Replacement is scheduled as required.
  • Includes plunger, seals, and check valve
  • System type: UPLC
  • Compatible with UPLC H-Class System, nanoUPLC System, 2D System
  • Compatible with UPLC ISM, UPLC BSM, UPLC QSM, UPLC H-Class QSM, nanoASM, nanoBSM
  • Product type: Performance maintenance kits
  • Units per package: 1 pk