Learn More
Waters Corp Waters Column, BEH, 45 Å, 1.7 μm, 4.6 mm x 150 mm
Catalog No. 50596921 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY APC AQ columns offer efficient size-based separations for water-soluble polymers. They feature patented Ethylene Bridge Hybrid (BEH) technology, providing a mechanically strong packed bed that resists shrinking and swelling, enabling easy mobile phase solvent switching.
  • Efficient size-based separations for water-soluble polymers.
  • Patented Ethylene Bridge Hybrid (BEH) technology.
  • Mechanically strong packed bed resists shrinking and swelling.
  • Facilitates easy mobile phase solvent switching.
  • Molecular weight range: 200 to 5,000 g/mole.
  • Delivers superior chromatographic efficiency, stability, and resolution.
  • Manufactured under stringent cGMP and ISO 9002 requirements.
Catalog No. 50-596-921 Supplier Waters Corp Supplier No. 186006975
ACQUITY APC AQ columns offer efficient size-based separations for water-soluble polymers. They feature patented Ethylene Bridge Hybrid (BEH) technology, providing a mechanically strong packed bed that resists shrinking and swelling, enabling easy mobile phase solvent switching.
  • Efficient size-based separations for water-soluble polymers.
  • Patented Ethylene Bridge Hybrid (BEH) technology.
  • Mechanically strong packed bed resists shrinking and swelling.
  • Facilitates easy mobile phase solvent switching.
  • Molecular weight range: 200 to 5,000 g/mole.
  • Delivers superior chromatographic efficiency, stability, and resolution.
  • Manufactured under stringent cGMP and ISO 9002 requirements.