Learn More
Waters Corp ACQUITY APC AQ - Aqueous Column, 450Å, 2.5 µm, 4.6 mm X 150 mm, 1/pk
Catalog No. NC1554434 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY APC AQ columns are designed to provide highly efficient size-based separations for water soluble polymer characterization. The sorbent uses patented Ethylene Bridge Hybrid (BEH) technology to provide a mechanical strong packed bed that resists shinking and swelling for easy mobile phase solvent switching. The expected molecular weight range for this column is approximatley 20,000 - 400, 000 g/mole.
Catalog No. NC1554434 Supplier Waters Corp Supplier No. 186006990
ACQUITY APC AQ columns are designed to provide highly efficient size-based separations for water soluble polymer characterization. The sorbent uses patented Ethylene Bridge Hybrid (BEH) technology to provide a mechanical strong packed bed that resists shinking and swelling for easy mobile phase solvent switching. The expected molecular weight range for this column is approximatley 20,000 - 400, 000 g/mole.