Learn More
Waters Corp ACQUITY APC Column, BEH, 125Å, 2.5 μm, 4.6 mm ID, 75 mm length
Supplier:  Waters Corp 186006978
Encompass_Preferred
ACQUITY APC AQ columns are designed to provide highly efficient size-based separations for water soluble polymer characterization. The sorbent uses patented Ethylene Bridge Hybrid (BEH) technology to provide a mechanically strong packed bed that resists shrinking and swelling for easy mobile phase solvent switching. The expected molecular weight range for this column is approximately 1,000 to 30,000 g/mole.
  • Ethylene Bridge Hybrid (BEH) technology
  • Mechanically strong packed bed
  • Resists shrinking and swelling for easy mobile phase solvent switching
  • Hybrid particle substrate
  • Spherical particle shape
  • 2.5 μm particle size
  • 125Å pore size
  • 4.6 mm inner diameter
  • 75 mm length
  • Expected molecular weight range of 1,000 to 30,000 g/mole
  • pH range of 1 to 8
Catalog No. 50-239-2127