Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp ACQUITY APC Column, 200 Å, 2.5 μm, 4.6 mm x 75 mm

Catalog No. 502392594 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This column is engineered for high-efficiency size-based separations, specifically for water-soluble polymer characterization. It utilizes patented Ethylene Bridge Hybrid (BEH) technology, resulting in a mechanically robust packed bed resistant to shrinking and swelling, which facilitates mobile phase solvent switching. The column is designed for an approximate molecular weight range of 3,000 to 70,000 g/mole.

  • Separation mode: SEC/GPC
  • Particle substrate: Hybrid
  • pH range: 1 to 8 pH
  • Silanol activity: Medium
  • Molecular weight range: 3,000 to 70,000
  • Particle shape: Spherical
  • Particle size: 2.5 μm
  • Endfitting type: Parker-style
  • Pore size: 200 Å
  • Inner diameter: 4.6 mm
  • Length: 75 mm
  • Particle technology: BEH
  • Technique: SEC
  • Units per package: 1

Catalog No. 50-239-2594 Supplier Waters Corp Supplier No. 186006983
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This column is engineered for high-efficiency size-based separations, specifically for water-soluble polymer characterization. It utilizes patented Ethylene Bridge Hybrid (BEH) technology, resulting in a mechanically robust packed bed resistant to shrinking and swelling, which facilitates mobile phase solvent switching. The column is designed for an approximate molecular weight range of 3,000 to 70,000 g/mole.

  • Separation mode: SEC/GPC
  • Particle substrate: Hybrid
  • pH range: 1 to 8 pH
  • Silanol activity: Medium
  • Molecular weight range: 3,000 to 70,000
  • Particle shape: Spherical
  • Particle size: 2.5 μm
  • Endfitting type: Parker-style
  • Pore size: 200 Å
  • Inner diameter: 4.6 mm
  • Length: 75 mm
  • Particle technology: BEH
  • Technique: SEC
  • Units per package: 1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.