Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, 200Å, 2.5 μm, 4.6 mm × 30 mm, 3K to 70K

Catalog No. 502392129 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

ACQUITY APC AQ columns are designed to provide highly efficient size-based separations for water-soluble polymer characterization. The sorbent uses patented Ethylene Bridge Hybrid (BEH) technology to provide a mechanically strong packed bed that resists shrinking and swelling for easy mobile phase solvent switching. The expected molecular weight range for this column is approximately 3,000 to 70,000 g/mole.

  • Separation mode: SEC/GPC
  • Particle substrate: Hybrid
  • PH range: 1 to 8
  • Endcapped: No
  • Silanol activity: Medium
  • Molecular weight range: 3,000 to 70,000 g/mole
  • Particle shape: Spherical
  • Particle size: 2.5 μm
  • Endfitting type: Parker-style
  • Pore size: 200 Å
  • System: APC
  • Particle technology: BEH
  • Technique: SEC
  • Inner diameter: 4.6 mm
  • Length: 30 mm
  • Units per package: 1 pk

Catalog No. 50-239-2129 Supplier Waters Corp Supplier No. 186006982
May include imposed supplier surcharges.
Add to Cart
Add to Cart

ACQUITY APC AQ columns are designed to provide highly efficient size-based separations for water-soluble polymer characterization. The sorbent uses patented Ethylene Bridge Hybrid (BEH) technology to provide a mechanically strong packed bed that resists shrinking and swelling for easy mobile phase solvent switching. The expected molecular weight range for this column is approximately 3,000 to 70,000 g/mole.

  • Separation mode: SEC/GPC
  • Particle substrate: Hybrid
  • PH range: 1 to 8
  • Endcapped: No
  • Silanol activity: Medium
  • Molecular weight range: 3,000 to 70,000 g/mole
  • Particle shape: Spherical
  • Particle size: 2.5 μm
  • Endfitting type: Parker-style
  • Pore size: 200 Å
  • System: APC
  • Particle technology: BEH
  • Technique: SEC
  • Inner diameter: 4.6 mm
  • Length: 30 mm
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.