Learn More
Waters Corp ACQUITY APC Column, BEH, 45Å, 1.7 μm, 4.6 mm
Supplier: Waters Corp 186006973
This column is engineered for highly efficient size-based separations, specifically for water-soluble polymer characterization. It utilizes patented Ethylene Bridge Hybrid (BEH) technology for a mechanically strong packed bed that resists shrinking and swelling, allowing for easy mobile phase solvent switching.
- Effective for characterizing molecules with a molecular weight range of approximately 200 to 5,000 g/mole.
- Designed for optimal separation performance with Advanced Polymer Chromatography™ System.
- Features an unbonded BEH substrate, recommended for aqueous solvent separations at room temperature (45°C).
- Manufactured in an ISO 9001-certified facility, ensuring stringent specifications for repeatable performance.
- Each column undergoes individual examination, and a Certificate of Batch Analysis is included on a connected eCord Intelligent Chip.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.