Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp ACQUITY APC Column, 450Å, 2.5 μm, 4.6 mm x 75 mm

Catalog No. 502391541 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

ACQUITY APC AQ columns are engineered for highly efficient size-based separations of water-soluble polymers. The sorbent uses patented Ethylene Bridge Hybrid (BEH) technology, providing a robust packed bed that resists shrinking and swelling, facilitating mobile phase solvent switching. They are suitable for molecular weights from 20,000 to 400,000 g/mole.

  • Employs Ethylene Bridge Hybrid (BEH) technology
  • Features a mechanically strong packed bed
  • Resists shrinking and swelling for easy mobile phase solvent switching
  • Operates in SEC/GPC separation mode
  • Composed of a hybrid particle substrate
  • Maintains stability across a pH range of 1 to 8
  • Utilizes spherical particle shapes
  • Features a particle size of 2.5 μm
  • Possesses a pore size of 450 Å
  • Has an inner diameter of 4.6 mm
  • Measures 75 mm in length

Catalog No. 50-239-1541 Supplier Waters Corp Supplier No. 186006988
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACQUITY APC AQ columns are engineered for highly efficient size-based separations of water-soluble polymers. The sorbent uses patented Ethylene Bridge Hybrid (BEH) technology, providing a robust packed bed that resists shrinking and swelling, facilitating mobile phase solvent switching. They are suitable for molecular weights from 20,000 to 400,000 g/mole.

  • Employs Ethylene Bridge Hybrid (BEH) technology
  • Features a mechanically strong packed bed
  • Resists shrinking and swelling for easy mobile phase solvent switching
  • Operates in SEC/GPC separation mode
  • Composed of a hybrid particle substrate
  • Maintains stability across a pH range of 1 to 8
  • Utilizes spherical particle shapes
  • Features a particle size of 2.5 μm
  • Possesses a pore size of 450 Å
  • Has an inner diameter of 4.6 mm
  • Measures 75 mm in length

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.