Learn More
Waters Corp Column, 900 Å, 2.5 μm, 4.6 mm × 150 mm
Supplier:  Waters Corp 186007251
Encompass_Preferred
ACQUITY APC AQ columns are designed to provide highly efficient size-based separations for water-soluble polymer characterization. The sorbent utilizes patented Ethylene Bridge Hybrid (BEH) technology, which offers a mechanically strong packed bed that resists shrinking and swelling, facilitating easy mobile phase solvent switching. The expected molecular weight range for this column is approximately 300,000 to 2,000,000 g/mole.
  • Efficient separations.
  • Ideal for characterizing water-soluble polymers.
  • Mechanically strong packed bed.
  • Resists shrinking and swelling, which allows for easy mobile phase solvent switching.
  • Broad molecular weight range of 300,000 to 2,000,000 g/mole.
Catalog No. 50-239-2461