Learn More
Waters Corp Calibration Kit, Polymethylmethacrylate, Middle MW, MS/UV/HRMS Detection
Supplier:  Waters Corp 186007544
Encompass_Preferred
APC Calibration Standards match the molecular weight range of ACQUITY APC XT Columns, eliminating manual preparation of custom mixes by providing targeted data points. Supplied in 12 x 32 mm amber glass vials with PTFE/silicone septa, cap color indicates molecular weight standards. Add solvent to dissolve and place in autosampler.
  • Eliminates manual preparation of custom calibration mixes.
  • Provides targeted data points for molecular weight range.
  • Supports MS, MS/MS, UV, HRMS detection.
  • Suitable for gel permeation applications.
  • Kit includes three sets of ten amber vials, each with 1.5 mg of polymethylmethacrylate (PMMA) standards.
  • PMMA standards cover molecular weights: 201K, 41.4K, 6.37K, 1.102K (red caps); 108K, 23.5K, 4.23K, 0.602K (light blue caps); 71.8K, 12.6K, 2.38K, 0.202K (white caps).
Catalog No. 50-239-2452