Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp ACQUITY APC Calibration Kit, Polymethylmethacrylate, Low MW

Catalog No. 502392453 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

APC™ Calibration Standards match ACQUITY APC™ XT Columns' molecular weight range, eliminating manual preparation. Supplied in 12 x 32 mm amber glass vials with PTFE/silicone septa, cap color indicates molecular weight standards. Users add solvent, dissolve standards, and place in autosampler per instructions.

  • Contains ten amber vials with green cap; containing 1.5 mg each of PMMA at Mp 12.6K, 4.23K, 1.102K
  • Contains ten amber vials with dark blue cap containing 1.5 mg each of PMMA at Mw 9.68K, 3.21K, 0.602K
  • Contains ten amber vials with black cap containing 1.5 mg each of PMMA at Mw 6.37K, 2.38K, 0.202K
  • Type: Calibration kit
  • Detection method: MS, MS/MS, UV, HRMS
  • UNSPSC: 41116107
  • Application: Gel permeation
  • Product type: Application kits

Catalog No. 50-239-2453 Supplier Waters Corp Supplier No. 186007543
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

APC™ Calibration Standards match ACQUITY APC™ XT Columns' molecular weight range, eliminating manual preparation. Supplied in 12 x 32 mm amber glass vials with PTFE/silicone septa, cap color indicates molecular weight standards. Users add solvent, dissolve standards, and place in autosampler per instructions.

  • Contains ten amber vials with green cap; containing 1.5 mg each of PMMA at Mp 12.6K, 4.23K, 1.102K
  • Contains ten amber vials with dark blue cap containing 1.5 mg each of PMMA at Mw 9.68K, 3.21K, 0.602K
  • Contains ten amber vials with black cap containing 1.5 mg each of PMMA at Mw 6.37K, 2.38K, 0.202K
  • Type: Calibration kit
  • Detection method: MS, MS/MS, UV, HRMS
  • UNSPSC: 41116107
  • Application: Gel permeation
  • Product type: Application kits

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.