Learn More
Waters Corp ACQUITY APC Calibration Kit, Polymethylmethacrylate, High MW
Supplier:  Waters Corp 186007545
Encompass_Preferred
APC™ Calibration Standards are designed for ACQUITY APC™ XT Columns, eliminating manual preparation of custom standard mixes. They provide accurate data points for targeted molecular weight ranges. Supplied in 12 x 32 mm amber glass vials with PTFE/silicone septa, cap color indicates the standards inside. Simply add solvent, dissolve, and place in autosampler.
  • Designed for XT Columns.
  • Eliminates manual custom standard mix preparation.
  • Ensures accurate data points for targeted molecular weight ranges.
  • Supplied in 12 x 32 mm amber glass vials with PTFE/silicone septa.
  • Cap color identifies molecular weight standards.
  • Easy autosampler integration after solvent addition and dissolution.
Catalog No. 50-239-2839