Learn More
Waters Corp Calibration Kit, Polystyrene, Middle MW, ACQUITY APC
Catalog No. 502390993 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY APC Polystyrene Middle MW Calibration Kit provides pre-packaged polystyrene standards for precise calibration in gel permeation chromatography. Optimized for ACQUITY APC XT Columns, it eliminates manual preparation of custom mixes, ensuring accurate calibration across a targeted middle molecular weight range.
  • Designed to match the molecular weight range of XT Columns.
  • Eliminates manual preparation of custom standard calibration mixes.
  • Provides the correct number of data points for the targeted molecular weight range.
  • Calibration standards come in 12 x 32 mm clear glass vials with PTFE/silicone septa.
  • Cap color indicates the molecular weight standards.
  • Easy to use: add solvent to dissolve standards, then place vial in autosampler.
  • Kit includes three sets of 10 clear vials (red, light blue, white caps), each with specific polystyrene standards.
Catalog No. 50-239-0993 Supplier Waters Corp Supplier No. 186007540
This ACQUITY APC Polystyrene Middle MW Calibration Kit provides pre-packaged polystyrene standards for precise calibration in gel permeation chromatography. Optimized for ACQUITY APC XT Columns, it eliminates manual preparation of custom mixes, ensuring accurate calibration across a targeted middle molecular weight range.
  • Designed to match the molecular weight range of XT Columns.
  • Eliminates manual preparation of custom standard calibration mixes.
  • Provides the correct number of data points for the targeted molecular weight range.
  • Calibration standards come in 12 x 32 mm clear glass vials with PTFE/silicone septa.
  • Cap color indicates the molecular weight standards.
  • Easy to use: add solvent to dissolve standards, then place vial in autosampler.
  • Kit includes three sets of 10 clear vials (red, light blue, white caps), each with specific polystyrene standards.