Learn More
Waters Corp Performance Maintenance Kit, ACQUITY APC Sample Manager -FTN, Annual, HPLC/UPLC
Catalog No. 502392335 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the ACQUITY APC Sample Manager – Flow Through Needle contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis.
  • Designed for HPLC and UPLC systems.
  • Intended for annual replacement.
  • Contains essential components for ACQUITY APC Sample Manager–FTN.
  • Includes syringe, standard needle, needle seat, injection cartridge, and filters.
Catalog No. 50-239-2335 Supplier Waters Corp Supplier No. 201000285
The Waters™ Performance Maintenance Kit for the ACQUITY APC Sample Manager – Flow Through Needle contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis.
  • Designed for HPLC and UPLC systems.
  • Intended for annual replacement.
  • Contains essential components for ACQUITY APC Sample Manager–FTN.
  • Includes syringe, standard needle, needle seat, injection cartridge, and filters.