Learn More
Waters Corp ACQUITY APC XT Column, 125Å, 2.5 μm, BEH Technology
Catalog No. 502392494 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY APC XT columns are engineered for highly efficient size-based separations of organic solvent soluble polymers. They utilize patented Ethylene Bridge Hybrid (BEH) technology, ensuring a mechanically robust packed bed that resists shrinking and swelling for easy mobile phase solvent switching. The column supports an approximate molecular weight range of 1,000 to 30,000 g/mole. All column documents, including COAs, are available online, reducing paper waste.
  • Efficient size-based separations of organic solvent soluble polymers.
  • Patented Ethylene Bridge Hybrid (BEH) technology for a strong, robust packed bed.
  • Resists shrinking and swelling for easy mobile phase solvent switching.
  • Supports molecular weight range of 1,000 to 30,000 g/mole.
  • Digital access to column documents, including COAs, for convenience and less waste.
Catalog No. 50-239-2494 Supplier Waters Corp Supplier No. 186006998
ACQUITY APC XT columns are engineered for highly efficient size-based separations of organic solvent soluble polymers. They utilize patented Ethylene Bridge Hybrid (BEH) technology, ensuring a mechanically robust packed bed that resists shrinking and swelling for easy mobile phase solvent switching. The column supports an approximate molecular weight range of 1,000 to 30,000 g/mole. All column documents, including COAs, are available online, reducing paper waste.
  • Efficient size-based separations of organic solvent soluble polymers.
  • Patented Ethylene Bridge Hybrid (BEH) technology for a strong, robust packed bed.
  • Resists shrinking and swelling for easy mobile phase solvent switching.
  • Supports molecular weight range of 1,000 to 30,000 g/mole.
  • Digital access to column documents, including COAs, for convenience and less waste.