Learn More
Waters Corp Column, 200Å, 2.5 μm, 4.6 x 30 mm
Supplier: Waters Corp 186007002
ACQUITY APC XT columns are designed for highly efficient size-based separations for organic solvent soluble polymer characterization, featuring Ethylene Bridge Hybrid (BEH) technology for a robust packed bed and a molecular weight range of 3,000 to 70,000 g/mole.
- Separation mode: SEC/GPC
- Particle substrate: Hybrid
- PH range: 1 pH to 12 pH
- Endcapped: No
- Silanol activity: Low
- Molecular weight range: 3,000 to 70,000
- Particle shape: Spherical
- Particle size: 2.5 μm
- Endfitting type: Parker-style
- Pore size: 200 Å
- Format: Column
- System: APC
- Particle technology: BEH
- Technique: GPC
- Inner diameter: 4.6 mm
- Length: 30 mm
- UNSPSC: 41115709
- Units per package: 1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.