Learn More
Waters Corp Column, 900 Å, 2.5 μm, 4.6 mm X 30 mm
Catalog No. 502391386 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY APC XT columns are optimized to provide highly efficient size-based separations for organic solvent soluble polymer characterization. The sorbent uses patented Ethylene Bridge Hybrid (BEH) technology to provide a mechanically strong packed bed that resists shrinking and swelling for easy mobile phase solvent switching.
  • Optimized to provide highly efficient size-based separations for organic solvent soluble polymer characterization.
  • Utilizes patented Ethylene Bridge Hybrid (BEH) technology.
  • Features a mechanically strong packed bed that resists shrinking and swelling.
  • Allows for easy mobile phase solvent switching.
  • Provides an expected molecular weight range of approximately 300,000 to 2,000,000 g/mole.
Catalog No. 50-239-1386 Supplier Waters Corp Supplier No. 186007252
ACQUITY APC XT columns are optimized to provide highly efficient size-based separations for organic solvent soluble polymer characterization. The sorbent uses patented Ethylene Bridge Hybrid (BEH) technology to provide a mechanically strong packed bed that resists shrinking and swelling for easy mobile phase solvent switching.
  • Optimized to provide highly efficient size-based separations for organic solvent soluble polymer characterization.
  • Utilizes patented Ethylene Bridge Hybrid (BEH) technology.
  • Features a mechanically strong packed bed that resists shrinking and swelling.
  • Allows for easy mobile phase solvent switching.
  • Provides an expected molecular weight range of approximately 300,000 to 2,000,000 g/mole.