Learn More
Waters Corp Waters ACQUITY APC XT Column, 125Å, 2.5 μm, 4.6 mm x 150 mm
Catalog No. NC1459404 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY APC XT columns are optimized for efficient size-based separations of organic solvent soluble polymers, utilizing patented Ethylene Bridge Hybrid (BEH) technology. The molecular weight range is approximately 1,000 to 30,000 g/mole.
  • Efficient size-based separations.
  • Patented BEH technology for strong packed bed.
  • Resists shrinking and swelling.
  • Superior chromatographic performance for polymer classes.
  • Extended temperature separations up to 90°C.
  • ISO 9001 facility manufacturing.
  • Individually tested.
Catalog No. NC1459404 Supplier Waters Corp Supplier No. 186007000
ACQUITY APC XT columns are optimized for efficient size-based separations of organic solvent soluble polymers, utilizing patented Ethylene Bridge Hybrid (BEH) technology. The molecular weight range is approximately 1,000 to 30,000 g/mole.
  • Efficient size-based separations.
  • Patented BEH technology for strong packed bed.
  • Resists shrinking and swelling.
  • Superior chromatographic performance for polymer classes.
  • Extended temperature separations up to 90°C.
  • ISO 9001 facility manufacturing.
  • Individually tested.