Learn More
Waters Corp ACQUITY Active Preheater, ACQUITY Premier, MaxPeak HPS, 12.5 in.
Catalog No. NC2541457 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Active Preheater is an essential component designed to enhance the performance of the ACQUITY Premier System. It incorporates MaxPeak High Performance Surfaces (HPS) technology to mitigate metal interactions, ensuring improved chromatographic results, especially for metal-sensitive analytes.
  • Compatible with ACQUITY Premier System.
  • Utilizes MaxPeak High Performance Surfaces (HPS) technology.
  • Minimizes adsorption losses caused by metal interactions.
  • Improves peak shape and reproducibility for metal-sensitive compounds.
  • Reduces the need for system passivation.
  • Features an inert sample flow path.
  • Includes active pre-heating for solvent conditioning.
  • Dimensions: 12.5 in.
  • Usage with 0.004 in. ID applications.
Catalog No. NC2541457 Supplier Waters Corp Supplier No. 205002234
This Active Preheater is an essential component designed to enhance the performance of the ACQUITY Premier System. It incorporates MaxPeak High Performance Surfaces (HPS) technology to mitigate metal interactions, ensuring improved chromatographic results, especially for metal-sensitive analytes.
  • Compatible with ACQUITY Premier System.
  • Utilizes MaxPeak High Performance Surfaces (HPS) technology.
  • Minimizes adsorption losses caused by metal interactions.
  • Improves peak shape and reproducibility for metal-sensitive compounds.
  • Reduces the need for system passivation.
  • Features an inert sample flow path.
  • Includes active pre-heating for solvent conditioning.
  • Dimensions: 12.5 in.
  • Usage with 0.004 in. ID applications.