Learn More
Waters Corp Performance Maintenance Kit, ACQUITY Arc Bio QSM-R, Mixers, Plungers, Seals
Catalog No. 502392769 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Performance Maintenance Kit contains all the Waters Quality Parts™ necessary to keep the ACQUITY Arc Bio Quaternary Solvent Manager operating at peak performance on an annual basis. It facilitates easy upkeep and regular scheduled maintenance, aiming to reduce downtime and prevent expensive repair costs.
  • Includes mixers, plungers, check valves, seals, and filters.
  • Designed for annual maintenance of the ACQUITY Arc Bio Quaternary Solvent Manager.
  • Undergoes strict quality control processes for proper performance.
Catalog No. 50-239-2769 Supplier Waters Corp Supplier No. 201000311
The Performance Maintenance Kit contains all the Waters Quality Parts™ necessary to keep the ACQUITY Arc Bio Quaternary Solvent Manager operating at peak performance on an annual basis. It facilitates easy upkeep and regular scheduled maintenance, aiming to reduce downtime and prevent expensive repair costs.
  • Includes mixers, plungers, check valves, seals, and filters.
  • Designed for annual maintenance of the ACQUITY Arc Bio Quaternary Solvent Manager.
  • Undergoes strict quality control processes for proper performance.