Learn More
Waters Corp Performance Maintenance Kit, ACQUITY Arc Bio SM-FTN, Syringe, Needle, Filters
Supplier:  Waters Corp 201000312
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the ACQUITY Arc Bio Sample Manager-Flow Through Needle contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis. The kit includes a syringe, standard needle, needle seat, injection cartridge, and filters.
  • Contains all necessary quality parts for annual maintenance.
  • Designed for ACQUITY Arc Bio Sample Manager-Flow Through Needle.
  • Includes syringe, standard needle, needle seat, injection cartridge, and filters.
Catalog No. 50-239-0704