Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Detector Solution, Liquid, 10 mL, System Performance, ACQUITY Arc

Catalog No. 502392690 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The ACQUITY Arc Refractive Index detector solution is designed for use with refractive index (RI) detectors within ACQUITY Arc systems. Its purpose is to evaluate the sensitivity, reproducibility, and linearity of the refractive index detector.

  • Separation mode: Reversed phase
  • Detection method: RI
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: LC
  • Instrument: ACQUITY Arc
  • Quantitative: Yes
  • Shelf life: 24 months
  • Shipping condition: Ambient
  • Volume: 10 mL each
  • Type: Reference
  • Application: System performance
  • Product type: Standards
  • Units per package: 1 pk
  • Contains hazardous material requiring special freight handling

Catalog No. 50-239-2690 Supplier Waters Corp Supplier No. 186008088
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The ACQUITY Arc Refractive Index detector solution is designed for use with refractive index (RI) detectors within ACQUITY Arc systems. Its purpose is to evaluate the sensitivity, reproducibility, and linearity of the refractive index detector.

  • Separation mode: Reversed phase
  • Detection method: RI
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: LC
  • Instrument: ACQUITY Arc
  • Quantitative: Yes
  • Shelf life: 24 months
  • Shipping condition: Ambient
  • Volume: 10 mL each
  • Type: Reference
  • Application: System performance
  • Product type: Standards
  • Units per package: 1 pk
  • Contains hazardous material requiring special freight handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.