Learn More
Waters Corp Standards, ACQUITY Arc, RI Detection, 10 mL, 24 Months Shelf Life
Catalog No. 502390675 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY Arc Refractive Index Start-up Standards Kit is intended to perform system precision and gradient performance tests on an ACQUITY Arc System with a RI detector.
  • Performs system precision and gradient performance tests.
  • Optimized for ACQUITY Arc system with a RI detector.
  • Uses reversed phase separation mode.
  • Features RI detection method.
  • Liquid formulation.
  • Stores at room temperature.
  • Compatible with LC system types.
  • Has a 24-month shelf life.
  • Ships under ambient conditions.
  • Comes in a 10 mL volume.
  • Reference type product.
  • Suitable for system calibration application.
  • Includes 1 unit per package.
Catalog No. 50-239-0675 Supplier Waters Corp Supplier No. 186008094
The ACQUITY Arc Refractive Index Start-up Standards Kit is intended to perform system precision and gradient performance tests on an ACQUITY Arc System with a RI detector.
  • Performs system precision and gradient performance tests.
  • Optimized for ACQUITY Arc system with a RI detector.
  • Uses reversed phase separation mode.
  • Features RI detection method.
  • Liquid formulation.
  • Stores at room temperature.
  • Compatible with LC system types.
  • Has a 24-month shelf life.
  • Ships under ambient conditions.
  • Comes in a 10 mL volume.
  • Reference type product.
  • Suitable for system calibration application.
  • Includes 1 unit per package.