Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp VanGuard Pre-column, C18, 1.7 μm, 300 Å, 2.1 mm × 5 mm

Catalog No. 50466015 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The ACQUITY UPLC Peptide BEH C18 VanGuard Pre-column is specifically designed and QC tested for peptide separations using cytochrome c digest standards. This pre-column utilizes BEH Technology C18, 300Å pore, 1.7μm particles and is engineered for routine use at pressures up to 15000 psi (1240 Bar) in ACQUITY UPLC System applications.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle size: 1.7 μm
  • Pore size: 300 Å
  • Inner diameter: 2.1 mm
  • Length: 5 mm
  • Application: Peptide
  • System compatibility: UHPLC, UPLC
  • Units per package: 3 pk

Catalog No. 50-466-015 Supplier Waters Corp Supplier No. 186004629
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The ACQUITY UPLC Peptide BEH C18 VanGuard Pre-column is specifically designed and QC tested for peptide separations using cytochrome c digest standards. This pre-column utilizes BEH Technology C18, 300Å pore, 1.7μm particles and is engineered for routine use at pressures up to 15000 psi (1240 Bar) in ACQUITY UPLC System applications.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle size: 1.7 μm
  • Pore size: 300 Å
  • Inner diameter: 2.1 mm
  • Length: 5 mm
  • Application: Peptide
  • System compatibility: UHPLC, UPLC
  • Units per package: 3 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.