Learn More
Waters Corp ACQUITY Premier Oligonucleotide BEH C18 Column, 300 Å, 1.7 μm, 2.1 x 100 mm
Catalog No. NC2895608 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY Premier Oligonucleotide BEH C18 Column, 300 Å, 1.7 μm, 2.1 x 100 mm with VanGuard FIT is used to separate synthetic oligonucleotides based on ion-pairing reversed-phase (IP-RP) chromatography. It is part of the MaxPeak Premier line of columns, which deliver an ultra-low non-adsorptive surface that maximizes sensitivity and reproducibility when separating metal-sensitive analytes, such as oligonucleotides.
  • QC tested using MassPrep OST standards to ensure repeatable performance.
  • Ultra-low non-adsorptive surface maximizes sensitivity and reproducibility for metal-sensitive analytes.
  • Integrated, replaceable 5 mm pre-column protects and extends analytical column life without affecting resolution.
  • VanGuard FIT cartridge replacements contain qualified sorbent and are easily replaced.
Catalog No. NC2895608 Supplier Waters Corp Supplier No. 186010755
This ACQUITY Premier Oligonucleotide BEH C18 Column, 300 Å, 1.7 μm, 2.1 x 100 mm with VanGuard FIT is used to separate synthetic oligonucleotides based on ion-pairing reversed-phase (IP-RP) chromatography. It is part of the MaxPeak Premier line of columns, which deliver an ultra-low non-adsorptive surface that maximizes sensitivity and reproducibility when separating metal-sensitive analytes, such as oligonucleotides.
  • QC tested using MassPrep OST standards to ensure repeatable performance.
  • Ultra-low non-adsorptive surface maximizes sensitivity and reproducibility for metal-sensitive analytes.
  • Integrated, replaceable 5 mm pre-column protects and extends analytical column life without affecting resolution.
  • VanGuard FIT cartridge replacements contain qualified sorbent and are easily replaced.