Learn More
Waters Corp UPLC Primary Check Valve Assembly, 2/pk
Catalog No. NC9352977 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The UPLC Primary Check Valve fits into the ACQUITY UPLC ACQUITY I-Class and the nanoACQUITY UPLC. The original part number was 700002704 and it was changed to p/n 700002596. This is a spring-loaded check valve.
Catalog No. NC9352977 Supplier Waters Corp Supplier No. 700002596
The UPLC Primary Check Valve fits into the ACQUITY UPLC ACQUITY I-Class and the nanoACQUITY UPLC. The original part number was 700002704 and it was changed to p/n 700002596. This is a spring-loaded check valve.