Learn More
Waters Corp Tubing Kit, 4 x 50 mm, Stainless Steel, for CM-A Column Managers
Catalog No. 502390604 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing kit contains stainless steel tubing assemblies designed to implement CM-A active column managers onto an ACQUITY On-line SPE System. It supports a four 50 mm column configuration. The kit can also be used to replace all stainless steel tubing in this configuration. Additionally, it includes the relevant Active Preheaters for a system configured with an ACQUITY PLUS CM-A.
  • Contains stainless steel tubing assemblies.
  • Designed for implementing CM-A active column managers onto an On-line SPE System.
  • Supports a four 50 mm column configuration.
  • Replaces all stainless steel tubing in this configuration.
  • Includes relevant Active Preheaters for systems configured with a PLUS CM-A.
Catalog No. 50-239-0604 Supplier Waters Corp Supplier No. 205001831
This tubing kit contains stainless steel tubing assemblies designed to implement CM-A active column managers onto an ACQUITY On-line SPE System. It supports a four 50 mm column configuration. The kit can also be used to replace all stainless steel tubing in this configuration. Additionally, it includes the relevant Active Preheaters for a system configured with an ACQUITY PLUS CM-A.
  • Contains stainless steel tubing assemblies.
  • Designed for implementing CM-A active column managers onto an On-line SPE System.
  • Supports a four 50 mm column configuration.
  • Replaces all stainless steel tubing in this configuration.
  • Includes relevant Active Preheaters for systems configured with a PLUS CM-A.