Learn More
Waters Corp Waters ACQUITY UPLC CSH C18 Column, 1.7 μm
Catalog No. 50791109 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY CSH C18 columns are designed to provide alternate selectivity compared to other reversed-phase UPLC columns. These ultra-efficient columns overcome peak shape asymmetry, poor loading capacity for basic compounds, column bleed, and rapid mobile-phase re-equilibration.
  • Provides superior peak shape for basic compounds, with low column bleed and high efficiency.
  • CSH technology offers trifluoroacetic acid (TFA) peak shape performance without the disadvantages of using TFA, allowing for improved performance with MS-friendly mobile phases like formic acid.
  • Enables faster and more straightforward method development.
  • Offers high loading of basic compounds under volatile, acidic, low pH mobile-phase conditions, reducing solvent consumption.
  • Features an intelligent chip for use on UPLC and Arc™ systems, ensuring consistent results.
Catalog No. 50-791-109 Supplier Waters Corp Supplier No. 176002140
ACQUITY CSH C18 columns are designed to provide alternate selectivity compared to other reversed-phase UPLC columns. These ultra-efficient columns overcome peak shape asymmetry, poor loading capacity for basic compounds, column bleed, and rapid mobile-phase re-equilibration.
  • Provides superior peak shape for basic compounds, with low column bleed and high efficiency.
  • CSH technology offers trifluoroacetic acid (TFA) peak shape performance without the disadvantages of using TFA, allowing for improved performance with MS-friendly mobile phases like formic acid.
  • Enables faster and more straightforward method development.
  • Offers high loading of basic compounds under volatile, acidic, low pH mobile-phase conditions, reducing solvent consumption.
  • Features an intelligent chip for use on UPLC and Arc™ systems, ensuring consistent results.