Learn More
Waters Corp Waters ACQUITY UPLC CSH C18 Column, 1.7 μm, 1 mm x 50 mm
Catalog No. 50791105 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed to offer alternate selectivity compared to other reversed-phase UPLC columns. They are ultra-efficient and address issues such as peak shape asymmetry, poor loading capacity for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 11
  • Maximum pressure of 18000 psi (1240 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 1.7 μm particle size
  • Parker-style endfitting type
  • 130 Å pore size
  • Column format
  • 185 surface area
  • Compatible with UPLC and UHPLC systems
  • CSH particle technology
  • L1 USP classification
  • 1 mm inner diameter
  • 50 mm length
  • 15% carbon load
  • Equipped with eCord
  • 3 units per package
Catalog No. 50-791-105 Supplier Waters Corp Supplier No. 176002136
These columns are designed to offer alternate selectivity compared to other reversed-phase UPLC columns. They are ultra-efficient and address issues such as peak shape asymmetry, poor loading capacity for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 11
  • Maximum pressure of 18000 psi (1240 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 1.7 μm particle size
  • Parker-style endfitting type
  • 130 Å pore size
  • Column format
  • 185 surface area
  • Compatible with UPLC and UHPLC systems
  • CSH particle technology
  • L1 USP classification
  • 1 mm inner diameter
  • 50 mm length
  • 15% carbon load
  • Equipped with eCord
  • 3 units per package