Learn More
Waters Corp Waters ACQUITY UPLC CSH C18 Column, 1.7 μm, 3 mm × 150 mm, 130 Å
Catalog No. 50791114 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY CSH C18 columns provide alternate selectivity in reversed-phase UPLC. Featuring Charged Surface Hybrid (CSH) Technology, they address peak shape asymmetry and enhance loading capacity for basic compounds, reduce column bleed, and enable rapid mobile-phase re-equilibration. This technology ensures superior selectivity and peak form in low-ionic-strength mobile phases, improving performance for basic compounds. UPLC Technology boosts resolution, sensitivity, and analysis speed.
  • Provides alternate selectivity for reversed-phase UPLC.
  • Overcomes peak shape asymmetry and poor loading capacity for basic compounds.
  • CSH technology delivers TFA peak shape performance without using TFA.
Catalog No. 50-791-114 Supplier Waters Corp Supplier No. 176002146
ACQUITY CSH C18 columns provide alternate selectivity in reversed-phase UPLC. Featuring Charged Surface Hybrid (CSH) Technology, they address peak shape asymmetry and enhance loading capacity for basic compounds, reduce column bleed, and enable rapid mobile-phase re-equilibration. This technology ensures superior selectivity and peak form in low-ionic-strength mobile phases, improving performance for basic compounds. UPLC Technology boosts resolution, sensitivity, and analysis speed.
  • Provides alternate selectivity for reversed-phase UPLC.
  • Overcomes peak shape asymmetry and poor loading capacity for basic compounds.
  • CSH technology delivers TFA peak shape performance without using TFA.