Learn More
Waters Corp Waters ACQUITY UPLC CSH C18 Columns, 1.7 μm, 3.0 × 100 mm
Catalog No. 50791113 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC CSH Columns are designed to provide superior peak shape for basic analytes when using formic acid or other low-ionic-strength, acidic mobile phases. These columns achieve this without the need for ion-pairing reagents.
  • Provides superior peak shape for basic analytes when using formic acid or other low-ionic-strength, acidic mobile phases.
  • Achieves results without the need for ion-pairing reagents due to a controlled, low-level, positive surface charge bonded to ethylene-bridged hybrid (BEH) particles.
  • Offers high efficiencies.
  • Provides excellent column stability for high-throughput method development.
Catalog No. 50-791-113 Supplier Waters Corp Supplier No. 176002145
ACQUITY UPLC CSH Columns are designed to provide superior peak shape for basic analytes when using formic acid or other low-ionic-strength, acidic mobile phases. These columns achieve this without the need for ion-pairing reagents.
  • Provides superior peak shape for basic analytes when using formic acid or other low-ionic-strength, acidic mobile phases.
  • Achieves results without the need for ion-pairing reagents due to a controlled, low-level, positive surface charge bonded to ethylene-bridged hybrid (BEH) particles.
  • Offers high efficiencies.
  • Provides excellent column stability for high-throughput method development.