Learn More
Waters Corp Waters Column, CSH C18, 130 Å, 1.7 μm, 1 mm x 50 mm
Supplier:  Waters Corp 186005292
Encompass_Preferred
ACQUITY CSH C18 columns offer alternative reversed-phase UPLC selectivity, resolving peak shape asymmetry, poor loading capacity, column bleed, and mobile-phase re-equilibration issues. These columns provide broad selectivity and high performance, with superior peak shape for basic compounds, low column bleed, and high efficiency, facilitating global method transfer.
  • Alternative reversed-phase selectivity.
  • Superior peak shape for basic compounds.
  • Overcomes peak asymmetry and column bleed.
  • CSH technology for TFA-free peak performance.
  • Supports MS-friendly mobile phases.
  • High loading capacity for basic chemicals.
  • pH range of 1–11 pH.
  • Pressure tolerance up to 18000 psi (1240 Bar).
Catalog No. 50-791-326