Learn More
Waters Corp Waters Corporation ACQUITY UPLC CSH C18 Column, 130Å, 1.7μm, 1 X 100 mm
Catalog No. 50791327 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY CSH C18 columns provide alternate selectivity in reversed-phase UPLC, addressing issues like peak shape asymmetry and column bleed. Utilizing Charged Surface Hybrid (CSH) Technology, they ensure excellent peak shape in low-ionic-strength mobile phases, benefiting basic compounds under low-pH conditions.
  • Offers alternate selectivity for reversed-phase separations.
  • Addresses peak shape asymmetry and column bleed.
  • Utilizes Charged Surface Hybrid (CSH) Technology for excellent peak shape.
  • Provides consistent and repeatable results for global method transfer.
  • Achieves TFA peak shape performance without using TFA.
  • Enables MS-friendly mobile phases and faster method development.
  • Delivers gains in resolution, sensitivity, and analysis speed.
  • Withstands working pressures up to 18000 psi (1240 Bar).
Catalog No. 50-791-327 Supplier Waters Corp Supplier No. 186005293
ACQUITY CSH C18 columns provide alternate selectivity in reversed-phase UPLC, addressing issues like peak shape asymmetry and column bleed. Utilizing Charged Surface Hybrid (CSH) Technology, they ensure excellent peak shape in low-ionic-strength mobile phases, benefiting basic compounds under low-pH conditions.
  • Offers alternate selectivity for reversed-phase separations.
  • Addresses peak shape asymmetry and column bleed.
  • Utilizes Charged Surface Hybrid (CSH) Technology for excellent peak shape.
  • Provides consistent and repeatable results for global method transfer.
  • Achieves TFA peak shape performance without using TFA.
  • Enables MS-friendly mobile phases and faster method development.
  • Delivers gains in resolution, sensitivity, and analysis speed.
  • Withstands working pressures up to 18000 psi (1240 Bar).