Learn More
Waters Corp ACQUITY UPLC CSH C18 Column, 130Å, 1.7 µm, 1 mm X 150 mm, 1/pk
Catalog No. 50791328 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY CSH C18 columns were designed to provide alternate selectivity compared to other reversed-phase UPLC columns. These ultra-efficient columns overcome peak shape asymmetry and poor loading capacity for basic compounds, column bleed, and rapid mobile-phase re-equilibration.
Catalog No. 50-791-328 Supplier Waters Corp Supplier No. 186005294
ACQUITY CSH C18 columns were designed to provide alternate selectivity compared to other reversed-phase UPLC columns. These ultra-efficient columns overcome peak shape asymmetry and poor loading capacity for basic compounds, column bleed, and rapid mobile-phase re-equilibration.