Learn More
Waters Corp Waters Corporation Column, 1.7 μm, 130 Å, 2.1 mm × 150 mm, 3/pk
Catalog No. 50791110 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY CSH C18 columns offer alternate selectivity for UPLC, addressing peak shape asymmetry and poor loading for basic compounds. They provide high loadability, enabling narrower columns, and ensure reproducible performance due to stringent manufacturing and testing.
  • Provides alternate selectivity for reversed-phase UPLC, overcoming peak shape asymmetry for basic compounds.
  • Offers high loading capacity, allowing narrower columns and reduced solvent and fraction volumes.
  • Ensures low column bleed, excellent batch-to-batch reproducibility, and superior peak shape for basic compounds.
  • Manufactured under cGMP, ISO 9001 certified conditions with ultra-pure reagents.
  • Individually tested with Performance Chromatogram and Certificate of Batch Analysis on eCord Intelligent Chip for reproducible performance.
Catalog No. 50-791-110 Supplier Waters Corp Supplier No. 176002142
ACQUITY CSH C18 columns offer alternate selectivity for UPLC, addressing peak shape asymmetry and poor loading for basic compounds. They provide high loadability, enabling narrower columns, and ensure reproducible performance due to stringent manufacturing and testing.
  • Provides alternate selectivity for reversed-phase UPLC, overcoming peak shape asymmetry for basic compounds.
  • Offers high loading capacity, allowing narrower columns and reduced solvent and fraction volumes.
  • Ensures low column bleed, excellent batch-to-batch reproducibility, and superior peak shape for basic compounds.
  • Manufactured under cGMP, ISO 9001 certified conditions with ultra-pure reagents.
  • Individually tested with Performance Chromatogram and Certificate of Batch Analysis on eCord Intelligent Chip for reproducible performance.