Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters ACQUITY UPLC CSH C18 Column, 1.7 μm, 3.0 × 30 mm

Catalog No. 50791111 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This ACQUITY UPLC CSH C18 column is designed for use with UPLC Systems, featuring sub-2-μm, fully porous Ethylene Bridged Hybrid (BEH) particles. It provides excellent peak shape for basic analytes when used with acidic mobile phases.

  • Utilizes sub-2-μm fully porous particle technologies.
  • Features Ethylene Bridged Hybrid (BEH) particles with a controlled, low-level, positive surface charge.
  • Provides excellent peak shape for basic analytes when used with formic acid or other low-ionic-strength acidic mobile phases.
  • Does not require ion-pairing reagents.
  • Has a particle size of 1.7 μm.
  • Dimensions of 3.0 × 30 mm.

Catalog No. 50-791-111 Supplier Waters Corp Supplier No. 176002143
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This ACQUITY UPLC CSH C18 column is designed for use with UPLC Systems, featuring sub-2-μm, fully porous Ethylene Bridged Hybrid (BEH) particles. It provides excellent peak shape for basic analytes when used with acidic mobile phases.

  • Utilizes sub-2-μm fully porous particle technologies.
  • Features Ethylene Bridged Hybrid (BEH) particles with a controlled, low-level, positive surface charge.
  • Provides excellent peak shape for basic analytes when used with formic acid or other low-ionic-strength acidic mobile phases.
  • Does not require ion-pairing reagents.
  • Has a particle size of 1.7 μm.
  • Dimensions of 3.0 × 30 mm.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.