Learn More
Waters Corp Waters Column Kit, CSH C18, 1.7 μm, 2.1 mm x 50 mm, 3 pk
Catalog No. 50791528 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit includes three CSH C18 columns for method validation, each with different sorbent batches. Using Charged Surface Hybrid (CSH) Technology, these columns offer enhanced selectivity, peak shape, and increased loading capacity for basic compounds in low ionic strength mobile phases.
  • Optimized for sub-2μm UPLC columns with HPLC method robustness.
  • Improves resolution, sensitivity, and analysis speed.
  • Increased loadability reduces solvent use and costs.
  • Manufactured to cGMP and ISO 9001 standards; individually tested.
  • eCord intelligent chip for data traceability.
  • C18 chemistry, reversed phase, hybrid particle substrate.
  • 1.7 μm particle size; 130 Å pore size.
  • pH range 1 to 11; max pressure 18000 psi.
  • Dimensions: 2.1 mm ID, 50 mm length.
  • 3 columns included for method validation.
Catalog No. 50-791-528 Supplier Waters Corp Supplier No. 186005571
This kit includes three CSH C18 columns for method validation, each with different sorbent batches. Using Charged Surface Hybrid (CSH) Technology, these columns offer enhanced selectivity, peak shape, and increased loading capacity for basic compounds in low ionic strength mobile phases.
  • Optimized for sub-2μm UPLC columns with HPLC method robustness.
  • Improves resolution, sensitivity, and analysis speed.
  • Increased loadability reduces solvent use and costs.
  • Manufactured to cGMP and ISO 9001 standards; individually tested.
  • eCord intelligent chip for data traceability.
  • C18 chemistry, reversed phase, hybrid particle substrate.
  • 1.7 μm particle size; 130 Å pore size.
  • pH range 1 to 11; max pressure 18000 psi.
  • Dimensions: 2.1 mm ID, 50 mm length.
  • 3 columns included for method validation.