Learn More
Waters Corp Waters ACQUITY UPLC CSH Fluoro-Phenyl Column, 1.7 μm, 1 mm, 50 mm
Supplier: Waters Corp 186005349
The ACQUITY CSH Fluoro-Phenyl column maximizes selectivity differences and provides method development freedom, particularly with low pH mobile phases. Its non-encapped sorbent enhances retention of acidic compounds.
- PFP/Fluoro-Phenyl chemistry, reversed phase separation.
- Hybrid, spherical particles with low silanol activity.
- pH range of 1 to 8.
- Maximum pressure of 18000 psi (1240 Bar).
- Particle size: 1.7 μm; Pore size: 130 Å.
- Dimensions: 1 mm inner diameter, 50 mm length.
- 10% carbon load; non-encapped.
- UPLC and UHPLC system compatibility.
- CSH particle technology for reproducibility and selectivity.
- Maintains peak shape and loading capacity for basic compounds without ion-pair reagents.
- Temperature limits: 60 °C at low pH, 45 °C at high pH.
- eCord enabled.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.